missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Bench-Top Biohazard Bag Holder

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

£19.50 - £79.95

Specifications

Autoclavable Autoclavable
Disposable Single-use
Width (English) 5 in.
Width (Metric) 12.7 cm
View More Specs

Products 2
Product Code Brand Product Type Includes Price Quantity & Availability  
Product Code Brand Product Type Includes Price Quantity & Availability  
13472509
View Documents Promotion Details
Bel-Art™
F13193-0102
Bag Holder Cover Snug-Fitting Plastic Cover Only
£19.50
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
15220828
View Documents Promotion Details
Bel-Art™
F13193-0500
Biohazard Bag Holder Kit Complete Kit: Bag Holder, Cover and Bags (100pk)
£79.95
1 set
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
 
Description

Description

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
Specifications

Specifications

Autoclavable
Single-use
5 in.
12.7 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Bench-Top Biohazard Bag Holder

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.