missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling

Product Code. 15529991
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Isotonic Saline
Machine Oil
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite (Bleach)
Toluene
Water
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Quantity:
2/Pk.
4/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position

  • Wide mouth 53mm (2 in.) for easy refilling
  • Translucent LDPE bottles with color-coded caps for easy identification
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve

  • Assortment is 4/Bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm

Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Red
Material LDPE
Quantity 2/Pk.
Chemical Label Acetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.