missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Isotonic Saline
Machine Oil
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite (Bleach)
Toluene
Water
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Quantity:
2/Pk.
4/Pk.
Description
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position
- Wide mouth 53mm (2 in.) for easy refilling
- Translucent LDPE bottles with color-coded caps for easy identification
- Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
- Methyl Ethyl Ketone and Toluene do not include vented valve
- Assortment is 4/Bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels
Specifications
Specifications
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Green |
Material | LDPE |
Quantity | 2/Pk. |
Chemical Label | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction