Learn More
Bel-Art™ SP Scienceware™ Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module
Brand: Bel-Art™ F42074-0020
16.80 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module
Automatically regenerates for thousands of cycles.
- Absorbs moisture from the air
- Inert, non-toxic, non-corrosive
- Won't disintegrate into powder even after repeated regeneration
- Color-changing moisture indicator turns from blue to light pink indicating saturation
- Pink/dark brown hues indicate replacement due
Specifications
Desiccant Replacement Bead | |
Silica Gel | |
Color-indicating silica gel beads for use in refilling the Secador™ auto-desiccator module | |
26 g |
Safety and Handling
- secador auto-desiccator module desicant replacement beads
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1
- Reproductive toxicity Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : cobalt dichloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.