Learn More
Bel Art™ Secador™ Desiccator Replacement Shelves
Product Code. 12388995 Shop All SP Scienceware Products
Click to view available options
Product Type:
Desiccator Shelf (Horizontal)
Desiccator Shelf (Vertical)
Unit Size:
Each
Product Code. 12388995
Brand: Bel Art™ F420740001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convert 4.0 units from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
  • Fit auto and nonelectric models
  • Durable, clear polystyrene construction
  • Perforated to faciilitate air flow
Specifications
Diameter (Metric) Outer 48 cm
Product Type Desiccator Shelf (Horizontal)
Material Polystyrene
For Use With (Application) Convert 4.0 units from vertical to horizontal desiccator type