missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Secador™ Gas-Purge Desiccator Cabinets

Available in a wide range of sizes for gas-purged desiccation

Brand:  Bel-Art™ F42074-1009

 View more versions of this product

Product Code. 15149124

  • £990.00 / Each
Estimated Shipment: 28-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available in a wide range of sizes for gas-purged desiccation

  • Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
  • Rear mounted gas port valves accept 1/4 in. tubing and can be closed to use standard desiccants
  • The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3L × 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down «oven style” door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3L × 13.4W × 8.4 in.H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in.H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in.H

Order Info

Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.

Specifications

Specifications

Not Autoclavable
Vertical
Left-hinged and gasketed
Rear mounted gas port valves, carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
3
1.87 cu. ft.
16.3 in.
13.4 in.
20.4 in.
16.3 in.
Amber
Copolyester
Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments
4.0
Chemicals, stains and fine cracks
16.3 x 13.4 in. (41.4 x 34.1 cm)
41.4 cm
34.1 cm
51.8 cm
41.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers