missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Secador™ Gas-Purge Desiccator Cabinets

Available in a wide range of sizes for gas-purged desiccation

Brand:  Bel-Art™ F42071-0002

 View more versions of this product

Product Code. 12776778

  • £372.00 / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available in a wide range of sizes for gas-purged desiccation

  • Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
  • Rear mounted gas port valves accept 1/4 in. tubing and can be closed to use standard desiccants
  • The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3L × 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down «oven style” door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3L × 13.4W × 8.4 in.H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in.H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in.H

Order Info

Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.

Specifications

Specifications

Not Autoclavable
Vertical
Left-hinged and gasketed
20 Cans
1.0
Chemicals, stains and fine cracks
16.3 x 13.4 in. (41.4 x 34.1 cm)
41.4 cm
8.9 in.
22.5 cm
4.5 in.
16.3 in.
Clear
Copolyester
Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments
Rear mounted gas port valves, carrying handles, latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer
1 fixed vented adjustable shelve
0.74 cu. ft.
16.3 in.
13.4 in.
34.1 cm
8.4 in.
21.3 cm
41.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.