Learn More
Bel-Art™ SP Scienceware™ Secador™ Gas-Purge Desiccator Cabinets
Available in a wide range of sizes for gas-purged desiccation
Brand: Bel-Art™ F42073-1009
Code : X3
Additional Details : Weight : 1.00000kg
Description
Available in a wide range of sizes for gas-purged desiccation
- Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
- Rear mounted gas port valves accept 1/4 in. tubing and can be closed to use standard desiccants
- The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3L × 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down «oven style” door
- U.S. Patents: 6,834,920; 7,318,630 and D474,626
Dimensions
- Secador 1.0: 0.74 cu. ft. internal volume, 16.3L × 13.4W × 8.4 in.H
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in.H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in.H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in.H
Order Info
Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.
Specifications
Not Autoclavable | |
Vertical | |
Left-hinged and gasketed | |
Rear mounted gas port valves, carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
3 | |
1.55 cu. ft. | |
16.3 in. | |
13.4 in. | |
16.4 in. | |
16.3 in. |
Amber | |
Copolyester | |
Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments | |
3.0 | |
Chemicals, stains and fine cracks | |
16.3 x 13.4 in. (41.4 x 34.1 cm) | |
41.4 cm | |
34.1 cm | |
41.7 cm | |
41.4 cm |