Learn More
SP Bel-Art™ Secador™ Standard Desiccator Cabinets
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
£501.00 - £1055.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Cabinet Style | Vertical |
Material | Copolyester |
Door Style | Left-hinged and gasketed |
For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant |
Product Code | Brand | Color | No. of Shelves | Volume (English) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Shelves | Volume (English) | Height (Metric) | Price | Quantity & Availability | ||
11822732
|
SP Bel-Art™
F42074-1007 |
Blue with Clear Door | 3 | 1.9 cu. ft. | 51.8 cm | N/A | |||
10551712
|
SP Bel-Art™
F42074-1006 |
Clear with Blue End Caps | 3 | 1.9 cu. ft. | 51.8 cm | N/A | |||
10260702
|
SP Bel-Art™
F42074-1000 |
Clear | 3 | 1.9 cu. ft. | 51.8 cm |
£1055.00
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
11816890
|
SP Bel-Art™
F42072-1008 |
Amber | 2 | 1.17 cu. ft. | 31.4 cm |
£513.76
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||
11826890
|
SP Bel-Art™
F42073-1008 |
Amber | 3 | 1.58 cu. ft. | 41.7 cm |
£643.09
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||
11836890
|
SP Bel-Art™
F42074-1008 |
Amber | 3 | 1.9 cu. ft. | 51.8 cm | N/A | |||
12318995
|
SP Bel-Art™
F42073-1000 |
Clear | 3 | 1.58 cu. ft. | 41.7 cm |
£752.00
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||
11842742
|
SP Bel-Art™
F42072-1000 |
Clear | 2 | 1.17 cu. ft. | 31.4 cm |
£501.00
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||
Description
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
- Can be used with desiccant cartridges, F42049-0100, F42046-0000
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. F42049-0100, F42046-0000
Specifications
Not Autoclavable | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant | |
Chemicals, stains and fine cracks | |
13.4 in. | |
16.3 in. |
Vertical | |
Left-hinged and gasketed | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
Removable Perforated | |
34.1 cm | |
41.4 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.