Learn More
Bel-Art™ SP Scienceware™ sidENTRY™ Clamping Ring Glove Box
Process samples wherever you need to. Small enough to move wherever it’s needed, the Bel-Art™ sidENTRY™ Glove Box features a large side entry door for placing material or equipment into the box.
Brand: Bel-Art™
2872.24 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 4.20000kg
Description
- Four adjustable levelers ensure glove box stability and leveling
- Gas purging of the glove box is possible via the supplied and easily assembled stopcock
- Hand entry through the 8” circular ports is customized to end user needs by selecting either the bellows type glove, economy sleeved glove, seamless sleeved glove or the iris port (H50025-0020), all sold separately
- Made of 6.35mm (¼”) clear acrylic
Order Info
Gloves sold separately.
Specifications
Acrylic | |
24 in. | |
2 | |
61 cm |
Processing samples or handling air sensitive materials | |
61 cm | |
24 in. |