Learn More
Bel-Art™ SP Scienceware™ Spinbox™ Spinplus™ Magnetic Stirring Bar Assortment
Useful assortments of popular sizes and shapes
£159.00 - £202.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | FDA, USP Class VI, ISO 9001:2008, SGS |
Material | Alnico Magnet |
Includes: Octagon figured 16 assorted bars, standard and micro; standard bars with pivot ring Octagon shaped 12 assorted bars, standard bars with pivot rings Round shaped 22 assorted bars with removable pivot rings on bars longer than 1 in. Metric Polygon shaped 18 assorted bars Color-coded, octagon shaped 24 assorted bars with pivot ring Color-coded micro bars 12 assorted bars Egg shaped bars 13 assorted bars Spinplus in. “+” shaped bars 5 assorted bars
Product Code | Brand | Shape | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Shape | Price | Quantity & Availability | |||||
11755064
|
Bel-Art™
F37185-0000 |
Octagon |
£202.00
Pack of 24 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
13203539
|
Bel-Art™
F37187-0000 |
Egg |
£159.00
Pack of 13 |
Estimated Shipment: 13-11-2024
Log in to see stock. |
|||||
Description
- Convenient storage box segregates sizes and styles for easy, no-rummage organization
- FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Available in octagon, egg and Spinplus™ shapes
- Autoclavable
Certifications
ISO 9001:2008
Specifications
Autoclavable | |
FDA, USP Class VI, ISO 9001:2008, SGS | |
Alnico Magnet |