Learn More
Bel-Art™ SP Scienceware™ Sterileware™ Disposable Sterile Sampler Knife
Polystyrene sampler knife featuring a blade that is serrated on one edge and smooth on the other
Brand:  Bel-Art™ H36965-0000
Product Code. 11336944
  • £348.00 / Pack of 200
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The serrated edge is capable of cutting seafood, meat, poultry, vegetables and other fibrous materials
  • Individually wrapped, sterilized and packed in a polyethylene (PE) plastic bag for one-time use and disposal
Specifications
Specifications
Polystyrene
Food sampling like Meats, Vegetables, Seafood and other Fibrous and Semi Solid Materials
20.9 cm
Sterile
Sampling Knife
8.25 in.
Single-use
Certificate of Processing for Sterilization