Learn More
Bel-Art™ SP Scienceware™ Sterileware™ Economy Sample Spoons
Molded high-impact polystyrene for durability and stiffness. SP Scienceware™ Sterileware™ Economy Sample Spoons are ideal for retrieving small, specific volume samples of powders or liquids.
Brand:  Bel-Art™ F36740-0025
 View more versions of this product
Product Code. 12391359
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
  • Flat-bottom shape permits use as a weighing boat
  • For one-time use
Order Info
100 individually sterile wrapped spoons or 12 nonsterile spoons per pack