Learn More
Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
Brand:  Bel-Art™ F13165-1419
 View more versions of this product
Product Code. 17662085
  • £254.00 / Pack of 200
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Specifications
Biohazard Disposal Bag
Red
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
2 mil
19 in.
14 in.
Autoclavable
Extra-strength, high-impact, high-temperature bags reduce autoclave time
Polymer
19 x 14 in. (48 x 36 cm)
48 cm
36 cm