Learn More
Bel-Art™ SP Scienceware™ Techni-Dome™ 360 Gas-Ported Desiccator
For contamination and dust-free desiccation, storage and degassing
Brand:  Bel-Art™ F42029-0001
Product Code. 12786758
  • £444.00 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Protects generous quantities of standards, samples, electronics or other moisture-sensitive items
  • Spacious 65 liter (4000 cu. in.) internal volume can hold items up to 45cm (18 in.) tall or wide, including racks, trays and instruments
  • Two gas ports are tapered and accept tubing with 1/4 to 3/8 in. I.D.; easy turning PTFE stopcocks facilitate turbulence-free entrance of gases, minimizing disturbance of contents
  • Polycarbonate construction is resistant to most common chemicals, as well as thermal stress; wipes clean easily
  • Includes two removable, polypropylene, 36.5cm (14.5 in.) and 22.5cm (9 in.) diameter shelves
  • Side handles provide easy portability and can be used as a place to affix tamper-evident seals
  • Gas ports can be closed to use desiccator with standard desiccant cartridges (08-594-17C, 08-594-17B, or 08-594-14B)
Specifications
Specifications
Transparent
Gas-ported Desiccator
65 L
18 in.
5 kg
Polycarbonate
2 gas ports, 2 removable polypropylene shelves (14.5 in. and 9 in.), polypropylene stopcock with PTFE plug and red handle
45 cm
11 lb.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote