missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Thistle Tube

Makes additions to burets and narrow neck containers easily

Brand:  Bel-Art™ F14751-0000

Product Code. 30193463

  • £62.40 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The flared top thistle tube is made from polypropylene which is chemical resistant, unbreakable and safe
  • Dimensions are 27mm (0.69 in.) I.D. top with a 41mm (0.53 in.) O.D. flare
  • Steam autoclavable at 121°C (250°F)
  • Pack of 12
Specifications

Specifications

41mm (OD); 27mm (ID)
Autoclavable
6.4mm
Polypropylene
Thistle tubes (Pack of 12)
Thistle Tube Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.