missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Three-In-One Reservoir
Shop All SP Scienceware Products
Click to view available options
:
Three-In-One Reservoir
Description
- A graduated single reagent reservoir with graduations at 10, 25, 40 and 50mL
- Reverse side has a reservoir with 8 separate 5mL (max. capacity) reagent channels and a reservoir with 12 separate 5mL (max. capacity) reagent channels
- Each compartment of the multi-channel reservoirs is numbered for reagent identification
- Suitable for use with all brands of multichannel pipettors
- Autoclavable and reusable
Specifications
Specifications
| Color | White |
| Capacity (Metric) | 60 mL |
| Material | Polypropylene |
| Sterility | Sterile |
| Type | Pipetting Reservoir |
| For Use With (Application) | Pipetting |
| Packaging | Bag |
| Quantity | 21 |
| Autoclavable | Autoclavable |
| Channels | 8 and 12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction