Learn More
Bel-Art™ SP Scienceware™ Tubing Sinkers
Excellent for use with pumps and dispensers, these TFE sinkers keep tubing at the bottom of a container
Brand:  Bel-Art™ F32903-0100
Product Code. 15544130
  • £87.25 / Pack of 2
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sinkers have hose barb ends for simple tubing connection
  • Solvent resistant
  • Autoclavable
Specifications
Specifications
Autoclavable
PTFE
Tubing Sinker
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Tubing Sinkers > Tubing Sinkers