missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Vibrasorb™ Economy Vibration Damping Mounts

Facilitate optimal use and help protect electromechanical and optical equipment from excessive wear and even structural damage that can be caused by exposure to vibration

Brand:  Bel-Art™ F18385-0000

Product Code. 11822742

  • £643.00 / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Absorbs vibrations down to 13Hz with maximum load of 16kg (35 lb.)
  • Four Vibro-Sorber™ cushions and neoprene feet support these 64mm (2.5 in.) thick platforms
  • High impact polystyrene shell
  • Smooth, non-slip neoprene working surface; easily wipes clean
  • Platform area: 34 x 27.5cm (13.5 x 11 in.)
  • Maximum carrying weight for effective vibration dampening: 11kg (24lb)

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.