missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Vikem™ Microscope Covers
Bel-Art™ SP Scienceware™ Vikem™ Microscope Covers are transparent vinyl covers that protect microscopes, balances and other instruments from dust and spills. Available in several sizes.
Brand: Bel-Art™ F24303-0000
This item is not returnable.
View return policy
Description
- Cover slides over the instrument when not in use
- Available in standard sizes
- Sizes (bottom x top x height): Large size: 16 x 11 x 18 in. (41 x 28 x 46cm) and Extra-large size: 19 x 15 x 22 in. (46 x 38 x 59cm)
Specifications
Microscope Cover | |
18 in. | |
16 in. | |
Transparent Vinyl | |
11 in. |
16 x 11 x 18 in. (41 x 28 x 46 cm) | |
46 cm | |
41 cm | |
L | |
28 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ SP Scienceware™ Vikem™ Microscope Covers > Microscope Covers, Size: Large
Spot an opportunity for improvement?Share a Content Correction