Learn More
Cytiva Benchkote sheet for ÄKTA systems
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
Brand:  Cytiva 2300-10094
38.02 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16497645
  • £40.05 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Why Benchkote sheet for lab ÄKTA systems?
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
  • Smooth, absorbent Whatman™ paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
  • Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
  • The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Absorbent Paper, Laminated Polyethylene
For use with ÄKTA go
25/Pk.