Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Protects the top buffer trays from buffer spillages and salt deposits
Brand: Cytiva 2300-10072
28.65 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.30000kg
Description
- Cover the top buffer tray to protect from buffer spillages and salt deposits
- Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
- Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
- Smooth, absorbent Whatman paper quickly absorbs spills
- Laminated polyethylene layer prevents flow-through to the working surface
- Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
- Convenient and easy to use
Specifications
Benchkote Sheets | |
Paper, Laminated Polyethylene | |
ÄKTA Avant | |
10/Pk. |