Learn More
Gibco™ BenchStable™ RPMI 1640
Gibco RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer.
£21.75 - £196.00
Specifications
Content And Storage | Store at room temperature. Protect from light. |
---|---|
Cell Line | HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16497060
|
Gibco™
A4192301 |
500 mL |
£21.75
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16407070
|
Gibco™
A4192302 |
10 x 500 mL |
£196.00
5000mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
BenchStable RPMI 1640 Medium has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional RPMI 1640 Medium when supplemented with ≥10% fetal bovine serum (FBS).
This RPMI 1640 Medium is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPES
Using BenchStable RPMI 1640 Medium
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L) and so requires a 5–10% CO2 environment to maintain physiological pH. RPMI 1640 Medium with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable RPMI 1640 Medium contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).
Specifications
Store at room temperature. Protect from light. | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
GlutaMAX, Phenol Red, Phenol Red | |
Energy efficient, Fewer resources used, Sustainable packaging | |
BenchStable™ |
HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas | |
1 X | |
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
Sterile-filtered | |
No HEPES, No Sodium Pyruvate | |
cGMP-compliant under the ISO 9001: 2015 standard | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.