missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Astell Scientific™ Benchtop Autoclaves
Totally self-contained autoclaves with heaters in chamber as standard, requiring no plumbing and only a single phase power supply.
Brand: Astell Scientific™ AMB440
Description
- Classic version requires manual water-fill and during the cycle condensate leaves the vessel via a silicone tube that can be connected to a heat-resistant bottle
- Media holdwarm and delayed start facility
- A 5.7 in. colour touchscreen with integrated USB data port
- Electropolished stainless steel chamber
- Simple pre-post vacuum option available
- Adjustable time and temperature
Specifications
13 A | |
63 L | |
600 mm | |
Benchtop Autoclave | |
230 V |
668 mm | |
346 mm | |
AMB | |
50/60 Hz | |
520 mm |