missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benchtop Coolers
GREENER_CHOICE

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.

Brand:  Thermo Scientific™ 5116-1600

247.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10253921

  • £404.00 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
TRUSTED_SUSTAINABILITY
Specifications

Specifications

190 mm
197 mm
140 mm
7.75 x 5.5 x 7.5 in. (197 x 140 x 190 mm)
3 x 4
Non-filled/clear lid
Benchtop Cooler
0.2 to 0.5 mL
0°C
7.5 in.
7.75 in.
5.5 in.
3x4 compartments, for 16-17mm dia.vials
<1°C, 5.0
Polycarbonate
Clear
12 Tubes (16 to 17 mm dia.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.