Learn More
Thermo Scientific™ Benchtop Dewar Flasks
GREENER_CHOICE
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.
Brand:  Thermo Scientific™ 4150-9000
694.29 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 10007910
  • £1110.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.
  • High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
  • Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
  • Use from -196°C to +100°C
  • Pour spout and bottom grip area for ease of removal of used liquids
  • Vented, insulated cover allows for vapor escape but maintains sample temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
338.14 fl oz.
1
396 mm
198mm
470mm
1
10 L
15.6 in.
7.8 in.
18.5 in.