missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEND3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23101-1-AP-150UL
This item is not returnable.
View return policy
Description
Product No. 23101-1-AP specifically recognizes 85 and 94 kDa isoforms.
Transcriptional repressor which associates with the NoRC (nucleolar remodeling complex) complex and plays a key role in repressing rDNA transcription. The sumoylated form modulates the stability of the NoRC complex component BAZ2A/TIP5 by controlling its USP21-mediated deubiquitination. Binds to unmethylated major satellite DNA and is involved in the recruitment of the Polycomb repressive complex 2 (PRC2) to major satellites ().Specifications
BEND3 | |
Polyclonal | |
Unconjugated | |
BEND3 | |
BEN domain containing 3, BEND3, KIAA1553, RP11 59I9.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57673 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5T5X7 | |
BEND3 | |
BEND3 Fusion Protein Ag19137 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BEND3 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction