Learn More
BEND3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23101-1-AP-20UL
 View more versions of this product
Product Code. 16842195
  • £135.00 / 20µL
Explore more special offers
Description
Description
Product No. 23101-1-AP specifically recognizes 85 and 94 kDa isoforms.
Transcriptional repressor which associates with the NoRC (nucleolar remodeling complex) complex and plays a key role in repressing rDNA transcription. The sumoylated form modulates the stability of the NoRC complex component BAZ2A/TIP5 by controlling its USP21-mediated deubiquitination. Binds to unmethylated major satellite DNA and is involved in the recruitment of the Polycomb repressive complex 2 (PRC2) to major satellites ().
Specifications
Specifications
BEND3
Polyclonal
Unconjugated
BEND3
BEN domain containing 3, BEND3, KIAA1553, RP11 59I9.2
Rabbit
Antigen Affinity Chromatography
RUO
57673
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5T5X7
BEND3
BEND3 Fusion Protein Ag19137
20 μL