missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendamustine hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Cytostatic agent that displays activity in non-Hodgkin's lymphomas. Exhibits bifunctionality; combines DNA alkylating properties with those of purine analogs.
Specifications
Quantity | 10 mg |
Formula Weight | 394.72 |
Percent Purity | >98% |
Chemical Name or Material | Bendamustine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'