missing translation for 'onlineSavingsMsg'
Learn More

Benfotiamine, MedChemExpress

MedChemExpress Benfotiamine (S-Benzoylthiamine O-monophosphate) is an analog of vitamin B1 with higher absorption and bioavailability than vitamin B1, and is commonly used as a food supplement for diabetic complications. Benfotiamine exhibits direct antioxidative capacity and prevents induction of DNA damage.

£185.13 - £711.48

Specifications

Chemical Name or Material Benfotiamine
CAS 22457-89-2
Color White
Purity Grade Notes Research
Molecular Formula C19H23N4O6PS
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16461669
Encompass_Preferred
View Documents
Medchem Express
HY-17374
1 g
£185.13
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16469817
Encompass_Preferred
View Documents
Medchem Express
HY-17374
5 g
£711.48
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Benfotiamine (S-Benzoylthiamine O-monophosphate) is an analog of vitamin B1 with higher absorption and bioavailability than vitamin B1, and is commonly used as a food supplement for diabetic complications. Benfotiamine exhibits direct antioxidative capacity and prevents induction of DNA damage.
Specifications

Specifications

Benfotiamine
White
C19H23N4O6PS
DMSO : ≥ 50 mg/mL (107.19 mM) ∣H2O : 0.67 mg/mL (1.44 mM; Need ultrasonic)
466.45
99.26%
Solid
22457-89-2
Research
S-Benzoylthiamine O-monophosphate
CC1=NC=C(CN(/C(C)=C(CCOP(O)(O)=O)/SC(C2=CC=CC=C2)=O)C=O)C(N)=N1
466.45
Research
Neuroscience-Neurodegeneration
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.