Learn More
Lenz Laborglasinstrumente™ Bent Adapter
Click to view available options
Standard Taper Joint:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 29/32 Upper Socket, 29/32 NS Lower Socket
Type:
Bent Cone Adapter
Bent Socket Adapter
Unit Size:
Each
Description
- Constructed from DURAN™ tubing
- With NS socket
- Bore: 2.5 mm
Specifications
Specifications
| Type | Bent Socket Adapter |
| Material | Duran Glass |
| Standard Taper Joint | NS 14/23 |