Learn More
Lenz Laborglasintrumente™ Bent Socket Adapter
Adapters with NS socket and bent glass olive (90°)
Brand:  Lenz Laborglasintrumente™ 05103003
Product Code. 11856124
  • £17.20 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 1 per pack
  • Made of DURAN™ tubing
Specifications
Specifications
NS 24/29
Adapter with NS socket and bent olive
Duran Tubing
1