missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde, 99.5+%, pure, redistilled, AcroSeal(R), Thermo Scientific Chemicals
38.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -26.0°C |
Density | 1.0430 g/mL |
Boiling Point | 179.0°C |
Flash Point | 64°C |
Assay Percent Range | 99.5+% |
Packaging | AcroSeal™ Glass bottle |
Molecular Formula | C7H6O |
Linear Formula | C6H5CHO |
Refractive Index | 1.5440 to 1.5460 |
Quantity | 100 mL |
Show More |
Safety and Handling
Product Identifier
- Benzaldehyde
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'