missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, 99.63%, MP Biomedicals™

£94.60 - £383.00

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
Synonym Alkyldimethylbenzylammonium chloride
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434612
View Documents
MP Biomedicals
0219015880
100 g
£94.60
100g
Please to purchase this item. Need a web account? Register with us today!
11454612
View Documents
MP Biomedicals
0219015891
1 kg
£383.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

8001-54-5
C9H13ClN
170.66
Alkyldimethylbenzylammonium chloride

Specifications

8001-54-5
Powder
C9H13ClN
170.66
White
0.9963
Alkyldimethylbenzylammonium chloride
Benzalkonium Chloride
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Benzalkonium Chloride

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4

Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.