missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzamidine hydrochloride hydrate, 98%, water ca 10-14%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 82°C to 85°C |
Quantity | 5 g |
Beilstein | 3594959 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
IUPAC Name | benzenecarboximidamide;hydrate;hydrochloride |
Formula Weight | 156.62 (Anhydrous) |
Percent Purity | 98% |
Assay | water ≈10 to 14% |
Chemical Name or Material | Benzamidine hydrochloride hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'