Learn More
Cytiva Benzamidine Sepharose™ 4 Fast Flow, High Sub Media
For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases and thrombin
Brand:  Cytiva 17-5123-10
Product Code. 11575015
  • £381.00 / 25mL
Explore more special offers
Description
Description
  • p-aminobenzamide (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity and allows easy scale-up
  • Binding capacity ≥35mg trypsin/mL media
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
90 μm
2 to 8 (long term); 1 to 9 (short term)
25 mL
Benzamine Sepharose 4 Fast Flow
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Benzamidine Sepharose™ 4 Fast Flow, High Sub Media > 25mL