missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone, 98%, Thermo Scientific Chemicals

£28.04 - £86.73

Chemical Identifiers

CAS 3562-84-3
Molecular Formula C17H12Br2O3
Molecular Weight (g/mol) 424.08
InChI Key WHQCHUCQKNIQEC-UHFFFAOYSA-N
Synonym benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
PubChem CID 2333
ChEBI CHEBI:3023
IUPAC Name (3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
SMILES CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15678695
View Documents Promotion Details
Thermo Scientific Acros
460900010
1 g
£28.04
1g
Please to purchase this item. Need a web account? Register with us today!
15688695
View Documents
Thermo Scientific Acros
460900050
5 g
£86.73
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3562-84-3
424.08
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
CHEBI:3023
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
C17H12Br2O3
WHQCHUCQKNIQEC-UHFFFAOYSA-N
2333
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone

Specifications

3562-84-3
0.5% max.
97.5% min. (GC)
C17H12Br2O3
Solubility in water: .. Other solubilities: soluble in hot methanol
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
424.08
CHEBI:3023
98%
152.0°C
Authentic
Glass Bottle
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
WHQCHUCQKNIQEC-UHFFFAOYSA-N
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
2333
424.08
Benzbromarone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 222-630-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only