missing translation for 'onlineSavingsMsg'
Learn More

Benzeneboronic acid, 98+%

Product Code. 11432397
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11432397

Brand: Thermo Scientific Alfa Aesar A14257.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid
SMILES OB(O)C1=CC=CC=C1

Specifications

Melting Point 214°C to 219°C
Quantity 50 g
Beilstein 970972
Sensitivity Hygroscopic
Merck Index 14,1068
Solubility Information Slightly soluble in water.
Formula Weight 121.93
Percent Purity ≥98%
Chemical Name or Material Benzeneboronic acid
compliance-icons
Product Identifier
  • Benzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : phenyl-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.