missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, 90%, technical, Thermo Scientific Chemicals

Product Code. 10724651
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-11-3, 7664-93-9
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.17
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color Yellow to Brown
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula C6H5SO3H
Quantity 1 kg
Beilstein 11,26
Merck Index 15,1071
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene,insoluble in ether and carbon disulfide
Formula Weight 158.17
Percent Purity 90%
Grade Technical
Physical Form Damp Crystalline Solid or Fused Mass
Chemical Name or Material Benzenesulfonic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.