missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10732012
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515-42-4
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.16
MDL Number MFCD00065179
InChI Key MZSDGDXXBZSFTG-UHFFFAOYSA-M
Synonym sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
PubChem CID 517327
IUPAC Name sodium;benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 450.0°C
Color White
Loss on Drying 2% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula SO3NaC6H5
Quantity 25 g
Merck Index 15,1071
Solubility Information (5% in water) Clear colorless to almost colorless
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonic acid, sodium salt, 98%
Show More Show Less
Product Identifier
  • Benzenesulfonic acid, sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.