Learn More
Benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
£24.10 - £126.00
Chemical Identifiers
CAS | 98-09-9 |
---|---|
Molecular Formula | C6H5ClO2S |
Molecular Weight (g/mol) | 176.614 |
MDL Number | MFCD00007426 |
InChI Key | CSKNSYBAZOQPLR-UHFFFAOYSA-N |
Synonym | phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride |
PubChem CID | 7369 |
IUPAC Name | benzenesulfonyl chloride |
SMILES | C1=CC=C(C=C1)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449822
|
Thermo Scientific Alfa Aesar
A10849.30 |
250 g |
£24.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439822
|
Thermo Scientific Alfa Aesar
A10849.0B |
1000 g |
£46.30
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459822
|
Thermo Scientific Alfa Aesar
A10849.0I |
5000 g | |||||||
Description
Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.
Solubility
Insoluble in water. Soluble in ether and alcohol.
Notes
Moisture sensitive. Incompatible with water, strong oxidizing agents, strong bases, methyl formamide and dimethyl sulfoxide.
Chemical Identifiers
98-09-9 | |
176.614 | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
7369 | |
C1=CC=C(C=C1)S(=O)(=O)Cl |
C6H5ClO2S | |
MFCD00007426 | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
benzenesulfonyl chloride |
Specifications
98-09-9 | |
1.377 | |
132°C (270°F) | |
C6H5ClO2S | |
MFCD00007426 | |
606926 | |
14,1072 | |
Insoluble in water. Soluble in ether and alcohol. | |
C1=CC=C(C=C1)S(=O)(=O)Cl | |
176.614 | |
176.62 | |
Benzenesulfonyl chloride |
14°C to 15°C | |
252°C | |
Irritating | |
1.5525 | |
UN2225 | |
Moisture sensitive | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
benzenesulfonyl chloride | |
7369 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BENZENE SULFONYL CHLORIDE
missing translation for 'einecsNumber' : 202-636-6
missing translation for 'rtecsNumber' : DB8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only