missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 10644882
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
IUPAC Name 1H-benzimidazole
SMILES N1C=NC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 174.0°C
Color Beige to Brown
Boiling Point >360.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 23, 131
Merck Index 15, 1083
Solubility Information Solubility in water: sparingly soluble. Other solubilities: more soluble in hot water,sparingly soluble in ether,practically insoluble in benzene and petroleum,soluble in aq. sol. of acids and strong alkalies,freely soluble in alcohol,boiling xylene
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzimidazole, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'