Learn More
Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals
£29.70 - £313.00
Chemical Identifiers
CAS | 496-41-3 |
---|---|
Molecular Formula | C9H6O3 |
Molecular Weight (g/mol) | 162.14 |
MDL Number | MFCD00005848 |
InChI Key | OFFSPAZVIVZPHU-UHFFFAOYSA-N |
Synonym | benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid |
PubChem CID | 10331 |
IUPAC Name | 1-benzofuran-2-carboxylic acid |
SMILES | OC(=O)C1=CC2=CC=CC=C2O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10032051
|
Thermo Scientific Acros
233260010 |
1 g | Glass bottle |
£29.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123401
|
Thermo Scientific Acros
233260050 |
5 g | Glass bottle |
£90.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10320904
|
Thermo Scientific Acros
233260250 |
25 g | Glass bottle |
£313.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
496-41-3 | |
162.14 | |
OFFSPAZVIVZPHU-UHFFFAOYSA-N | |
10331 | |
OC(=O)C1=CC2=CC=CC=C2O1 |
C9H6O3 | |
MFCD00005848 | |
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid | |
1-benzofuran-2-carboxylic acid |
Specifications
496-41-3 | |
100.0 | |
White to Yellow | |
98% | |
MFCD00005848 | |
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid | |
OFFSPAZVIVZPHU-UHFFFAOYSA-N | |
1-benzofuran-2-carboxylic acid | |
10331 | |
98% | |
Benzo[b]furan-2-carboxylic acid |
97.5 | |
195.0°C to 196.0°C | |
Authentic | |
C9H6O3 | |
15, 2549 | |
Solubility in water: partially soluble in water,soluble in boiling wat. Other solubilities: good soluble in acetone and ethanol,slightly soluble in chloroform,carbon disulfide | |
OC(=O)C1=CC2=CC=CC=C2O1 | |
162.14 | |
162.14 | |
Crystals or Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-818-9
- Benzo¢b!furan-2-carboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only