missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-carbonyl chloride, 97%, Thermo Scientific™

£47.70 - £133.00

Chemical Identifiers

CAS 111964-21-7
Molecular Formula C9H5ClO2
Molecular Weight (g/mol) 180.587
MDL Number MFCD11841066
InChI Key YUHWSRVMUQSCNL-UHFFFAOYSA-N
Synonym benzo b furan-3-carbonyl chloride, 3-benzofurancarbonylchloride, benzofuran-3-carbonyl chloride, benzofuran-3-carboxylic acid chloride, acmc-1cgrg
PubChem CID 13024720
IUPAC Name 1-benzofuran-3-carbonyl chloride
SMILES C1=CC=C2C(=C1)C(=CO2)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10193504
View Documents
Thermo Scientific Maybridge
CC27502CB
250 mg Amber Glass Bottle
£47.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10103514
View Documents
Thermo Scientific Maybridge
CC27502DA
1 g Amber Glass Bottle
£133.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

111964-21-7
180.587
YUHWSRVMUQSCNL-UHFFFAOYSA-N
13024720
C1=CC=C2C(=C1)C(=CO2)C(=O)Cl
C9H5ClO2
MFCD11841066
benzo b furan-3-carbonyl chloride, 3-benzofurancarbonylchloride, benzofuran-3-carbonyl chloride, benzofuran-3-carboxylic acid chloride, acmc-1cgrg
1-benzofuran-3-carbonyl chloride

Specifications

111964-21-7
100.0
C9H5ClO2
benzo b furan-3-carbonyl chloride, 3-benzofurancarbonylchloride, benzofuran-3-carbonyl chloride, benzofuran-3-carboxylic acid chloride, acmc-1cgrg
C1=CC=C2C(=C1)C(=CO2)C(=O)Cl
180.587
180.59
Benzo[b]furan-3-carbonyl chloride
96.5
97%
MFCD11841066
YUHWSRVMUQSCNL-UHFFFAOYSA-N
1-benzofuran-3-carbonyl chloride
13024720
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications