missing translation for 'onlineSavingsMsg'
Learn More

Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester, 97%

Product Code. 10737728 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10737728

Brand: Thermo Scientific Acros 430570010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1168135-03-2 | C12H15BN2O2S | 262.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1168135-03-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 84.0°C to 86.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H15BN2O2S
MDL Number MFCD11867874
Quantity 1 g
Synonym benzo c 1,2,5 thiadiazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo c 1,2,5 thiadiazole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole-5-boronic acid, pinacol ester, benzo c 1,2,5 thiadiazol-5-ylboronic acid pinacol ester, amtb346, benzo c 1,2,5 thiadiazole-5-boronic acid, pinacol ester
InChI Key KISHNZJGTMYYKH-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NSN=C2C=C1
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol) 262.13
PubChem CID 46738015
Formula Weight 262.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • Benzo¢c!¢1,2,5!thiadiazole-5-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.