missing translation for 'onlineSavingsMsg'
Learn More
Benzofuroxan, 98%, Thermo Scientific Chemicals Product Code.: 11378255

Benzofuroxan, 98%, Thermo Scientific Chemicals

Product Code. 11378255
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11378255

Brand: Thermo Scientific Alfa Aesar L10144.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-96-6
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005807
InChI Key OKEAMBAZBICIFP-UHFFFAOYSA-N
Synonym benzofuroxan, benzofuroxane, benzofurazan 1-oxide, benzofurazan oxide, 2,1,3-benzoxadiazole 1-oxide, 2,1,3-benzoxadiazole, 1-oxide, benzofurazan, 1-oxide, 2,1,3-benzoxadiazol-1-ium-1-olate, benzo c 1,2,5-oxadiazol-1-ol, benzofurazan n-oxide
PubChem CID 68075
IUPAC Name 3-oxido-2,1,3-benzoxadiazol-3-ium
SMILES C1=CC2=NO[N+](=C2C=C1)[O-]

Specifications

Melting Point 67°C to 71°C
Quantity 50 g
Beilstein 125112
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 136.11
Percent Purity 98%
Chemical Name or Material Benzofuroxan

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10115852 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL10144.18,L10144.09
Lot NumberY17H024 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL10144.18,L10144.09
Lot NumberU25J014 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberL10144.18,L10144.09
Lot NumberU02I025 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberL10144.18,L10144.09
Lot NumberJ22U025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberL10144.18,L10144.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzofuroxan, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.