missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic Acid, 99.5+%, Extra Pure, SLR, Fisher Chemical™
£78.65 - £108.00
Description
Benzoic Acid, 99.5+%, Extra Pure, SLR
Chemical Identifiers
65-85-0 | |
C7H6O2 | |
2398 |
Specifications
65-85-0 | |
White | |
249°C | |
C7H6O2 | |
122.12g/mol |
121°C | |
2.5 to 3.5 | |
Plastic powder jar | |
2398 | |
Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Benzoic acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.