Learn More
Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals
Description
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
pH | 2.5 to 3.5 |
Melting Point | 121°C to 124°C |
Boiling Point | 249°C |
Color | White |
Beilstein | 636131 |
Merck Index | 14,1091 |
Solubility Information | Soluble in acetone,benzene,chloroform,alcohol,diethyl ether,hexane and liquid ammonia. Slightly soluble in water. |
Formula Weight | 122.12 |
Percent Purity | ≤99.5% |
Grade | ACS |
Show More |
RUO – Research Use Only