Learn More
Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals
£25.73 - £96.58
Chemical Identifiers
CAS | 65-85-0 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.123 |
MDL Number | MFCD00002398 |
InChI Key | WPYMKLBDIGXBTP-UHFFFAOYSA-N |
Synonym | benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv |
PubChem CID | 243 |
ChEBI | CHEBI:30746 |
IUPAC Name | benzoic acid |
SMILES | C1=CC=C(C=C1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333288
|
Thermo Scientific Alfa Aesar
036230.14 |
25 g |
£25.73
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323288
|
Thermo Scientific Alfa Aesar
036230.22 |
100 g |
£32.46
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343288
|
Thermo Scientific Alfa Aesar
036230.36 |
500 g |
£96.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65-85-0 | |
122.123 | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
243 | |
benzoic acid |
C7H6O2 | |
MFCD00002398 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
CHEBI:30746 | |
C1=CC=C(C=C1)C(=O)O |
Specifications
2.5 to 3.5 | |
121°C to 124°C | |
White | |
MFCD00002398 | |
14,1091 | |
Soluble in acetone,benzene,chloroform,alcohol,diethyl ether,hexane and liquid ammonia. Slightly soluble in water. | |
C1=CC=C(C=C1)C(=O)O | |
122.123 | |
CHEBI:30746 | |
≤99.5% | |
1.32 g/mL | |
Weak, characteristic | |
Benzoic acid |
65-85-0 | |
249°C | |
C7H6O2 | |
636131 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
benzoic acid | |
243 | |
122.12 | |
ACS | |
121°C (250°F) | |
Crystalline |
Safety and Handling
GHS H Statement
H318-H302-H335
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
P260-P264b-P270-P280-P302+P352-P305+P351+P338-P310-P314-P332+P313-P362-P501c
H315-H318-H372-H500
missing translation for 'einecsNumber' : 200-618-2
missing translation for 'rtecsNumber' : DG0875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Benzoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Benzoic acid
RUO – Research Use Only