missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic acid, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Appearance | Complying |
Percent Purity | 99.9% (min.) |
Grade | Reagent Pharmacopeia Acids |
Quantity | 100 g |
Packaging | Plastic Bottle |
Physical Form | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'